Learn More
Invitrogen™ Ibalizumab Recombinant Monoclonal Antibody
Artikelnummer. 30282599
Change view
Click to view available options
Menge:
1 mg
100 μg
Packungsgröße:
1 Milligramm
100 Mikrogramm
Artikelnummer. Menge unitSize
30282599 1 mg 1 Milligramm
30283535 100 μg 100 Mikrogramm
2 options
Artikelnummer. 30282599
Marke: Invitrogen™ MA559218
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Recombinant Monoclonal Antibody
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Ibalizumab (also known asibalizumab-uiykand formerly known as TNX-355) is a monoclonal antibody that binds to CD4 receptors on the surface of CD4-positive cells, preventing HIV particle entry into the lymphocytes. It is an advanced and current antibody in development for the treatment of HIV/AIDS. It has been developed by Taimed biologics and Theratechnologies 8, 5.
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen Ibalizumab Humanized
Anwendungen ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Klassifikation Recombinant Monoclonal
Konzentration 1 mg/mL
Konjugat Unconjugated
Zusammensetzung 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Gen-Alias Hu5A8; TMB-355; TNX-355
Wirtsspezies Human
Reinigungsverfahren Protein A
Menge 1 mg
Regulatorischer Status RUO
Primär oder sekundär Primary
Zielspezies Human
Inhalt und Lagerung -20°C, Avoid Freeze/Thaw Cycles
Produkttyp Antibody
Form Lyophilized
Isotype IgG4SP