missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ICAM-1 Human ELISA Development Kit (ABTS), PeproTech®, PeproTech®
Click to view available options
Menge:
2 x 96 Tests
Packungsgröße:
2 Stück
Beschreibung
ICAM-1 (CD54) is an 85-110 kDa single-chain type 1 integral membrane glycoprotein with an extracellular domain of five immunoglobulin superfamily repeats, a transmembrane region and a cytoplasmic domain. ICAM-1 has 7 potential N-linked glycosylation sites and shares considerable amino acid sequence homology with ICAM-3 (CD50) and ICAM-2 (CD102). ICAM-1 binds to integrins of type CD11a/CD18 (leukocyte adhesion molecule, LFA-1), or CD11b/CD18 (Mac-1) and is exploited by Rhinovirus as a receptor. ICAM-1 is expressed by activated endothelial cells and detected on epithelial cells, fibroblasts, chondrocytes, B lymphocytes, T lymphocytes (low), monocytes, macrophages, dendritic cells and neutrophils, with lower levels that increase upon inflammation. ICAM-1 is also detected in some carcinoma and melanoma cells. Soluble ICAM-1 is detectable in the plasma and is elevated in patients with various inflammatory syndromes.
Spezifikation
Spezifikation
| Zugriffsnummer | P05362 |
| Produkttyp | ELISA |
| Gen-ID (Entrez) | 3383 |
| Gensymbol | ICAM1 |
| Menge | 2 x 96 Tests |
| Kennzeichnung | RUO |
| Lagerungsbedingungen | Informationen zur komponentenspezifischen Lagertemperatur finden Sie in der Produktdokumentation. |
| Ziel | Human |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur