Learn More
Invitrogen™ ICOS (CD278) Recombinant Rabbit Monoclonal Antibody (8P2G3)
Rabbit Recombinant Monoclonal Antibody
Marke:  Invitrogen™ MA558129
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 30282730
  • 185.00 € / 20 Mikroliter
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Immunogen sequence: EINGSADHRM FSFHNGGVQI SCKYPETVQQ LKMRLFRERE VLCELTKTKG SGNAVSIKNP MLCLYHLSNN SVSFFLNNPD SSQGSYYFCS LSIFDPPPFQ ERNLSGGYLH IYESQLCCQL KL.
ICOS (CD278) encodes a protein that is a member of the CD28 and CTLA-4 cell-surface receptor family. It functions as a homodimer and has a crucial role in cell-cell signaling, immune responses, and the regulation of cell proliferation. ICOS has been linked to various disorders, including Immunodeficiency, Common Variable, 1 and Common Variable Immunodeficiency.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
ICOS (CD278)
Recombinant Monoclonal
1.63 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
Q9R1T7, Q9WVS0
ICOS
Recombinant fusion protein containing a sequence corresponding to amino acids 21-142 of mouse ICOS/CD278 (NP_059508.2).
20 μL
Primary
Mouse, Rat
Antibody
IgG
ELISA, Western Blot
8P2G3
Unconjugated
ICOS
Activation-inducible lymphocyte immunomediatory molecule; Ailim; CCLP; CD278; CD28 and CTLA-4-like protein; CD28-related protein 1; CRP-1; CVID1; H4; Icos; inducible costimulator; inducible T cell costimulator; inducible T cell co-stimulator; inducible T-cell costimulator; inducible T-cell co-stimulator; Ly115
Rabbit
Protein A
RUO
54167, 64545
-20°C, Avoid Freeze/Thaw Cycles