Learn More
Tocris Bioscience™ IDRA 21
Inhibits AMPA receptor desensitization
Beschreibung
Inhibits AMPA receptor desensitization and enhances cognition by a related mechanism. More able to cross the blood-brain barrier than cyclothiazide (Cat. No. 0713).Spezifikation
| >98% |