missing translation for 'onlineSavingsMsg'
Learn More

IFN-alpha/beta R1 Antibody (Medarex patent anti-IFNAR-1) [Janelia Fluor™ 585], Novus Biologicals™

Artikelnummer. 30168127
Change view
Click to view available options
Menge:
0.1 mL
Unit Size:
0.10 Milliliter
Product Code. Menge unitSize
30168127 0.1 mL 0.10 Milliliter
1 options
This item is not returnable. View return policy

Product Code. 30168127

Brand: Novus Biologicals (Bio-Techne) NBP328805JF585

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Human Monoclonal Antibody

IFN-alpha/beta R1 Monoclonal antibody specifically detects IFN-alpha/beta R1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN-alpha/beta R1
Anwendungen Flow Cytometry, ELISA, Functional Assay
Klassifikation Monoclonal
Klon Medarex patent anti-IFNAR-1
Konjugat Janelia Fluor 585
Zusammensetzung 50mM Sodium Borate
Gen-Alias alpha-type antiviral protein, AVP, beta-type antiviral protein, CRF2-1, Cytokine receptor class-II member 1, Cytokine receptor family 2 member 1, human interferon-alpha receptor (HuIFN-alpha-Rec)10IFRC, IFN-alpha/beta receptor 1, IFN-alpha-REC, IFNAR, IFNBR, IFN-R-1, interferon (alpha, beta and omega) receptor 1, interferon alpha/beta receptor 1, interferon-alpha/beta receptor alpha chain, interferon-beta receptor 1, Type I interferon receptor 1
Wirtsspezies Human
Immunogen IFNAR1
Reinigungsverfahren Protein A purified
Menge 0.1 mL
Regulatorischer Status RUO
Forschungsgebiet Neuroscience, Phospho Specific
Primär oder sekundär Primary
Gen-ID (Entrez) 3454
Zielspezies Human
Inhalt und Lagerung Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.