missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IFN beta Recombinant Rabbit Monoclonal Antibody (527)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA560290
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
IFN beta Recombinant Monoclonal Antibody for ELISA
Type I Interferons (IFN-alpha/beta) are produced primarily in response to viral infection by Natural IFN-producing cells (IPCs) as part of the host immune response. IFNs can also inhibit the development of tumors. IFN-beta binding results in the activation of the tyrosine kinases Jak1 and Tyk2, phosphorylation of members of the STAT family of transcription factors, and the transcription and expression of the immune response genes. More recently, several members of the toll-like receptor (TLR) family were found to stimulate the production IFN-beta. IFN-beta is currently used clinically for treatment of tumors, infections and multiple sclerosis.
Spezifikation
| IFN beta | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P01574 | |
| Ifnb1 | |
| Recombinant Human IFN-beta protein (Met1-Asn187). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA | |
| 527 | |
| Unconjugated | |
| Ifnb1 | |
| Fibroblast interferon; Ifb; IFF; IFN b; IFN β; IFNB; Ifnb1; IFNbeta; IFN-beta; IFNβ; Interferon; Interferon b; Interferon beta; interferon beta 1; interferon beta 1, fibroblast; interferon beta protein; interferon, beta 1; interferon, beta 1, fibroblast; interferon-beta; RP11-113D19.1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3456 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur