missing translation for 'onlineSavingsMsg'
Learn More

IFN-omega/IFNW1 Antibody, Novus Biologicals™

Artikelnummer. 18619775 Shop All Bio Techne Products
Change view
Click to view available options
Menge:
100 μg
50 μg
Unit Size:
100 Mikroliter
50 Mikroliter
Product Code. Menge unitSize
18619775 50 μg 50 Mikroliter
18621896 100 μg 100 Mikroliter
2 options
This item is not returnable. View return policy

Product Code. 18619775

Brand: Novus Biologicals NBP29925650ul

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IFN-omega/IFNW1 Polyclonal antibody specifically detects IFN-omega/IFNW1 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN-omega/IFNW1
Anwendungen ELISA
Klassifikation Polyclonal
Konjugat Unconjugated
Verdünnung ELISA 1:5000-1:10000
Zusammensetzung 0.2 um filtered solution in PBS
Gen-Alias IFN-omega 1, interferon omega-1, Interferon alpha-II-1, interferon omega-1, interferon, omega 1
Wirtsspezies Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human IFN-omega/IFNW1 (Accession#: P05000; Met1-Ser195)
Reinigungsverfahren Antigen and protein A Affinity-purified
Menge 50 μg
Regulatorischer Status RUO
Forschungsgebiet Cell Cycle and Replication
Primär oder sekundär Primary
Gen-ID (Entrez) 3467
Zielspezies Human
Inhalt und Lagerung Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.