Learn More
Invitrogen™ Mouse IgA Monoclonal Antibody (11-44-2), PE, eBioscience™
Beschreibung
Description: The 11-44-2 monoclonal antibody reacts with the alpha heavy chain of mouse IgA. It does not react with other classes of mouse immunoglobulin including IgD, IgG or IgM. IgA is expressed as a membrane-bound form as well as a soluble dimer. Fluorochrome conjugated 11-44-2 is used as a detection secondary for mouse IgA. Applications Reported: 11-44-2 has been reported for use in flow cytometric analysis to detect the surface IgA on mouse B cells. Applications Tested: This 11-44-2 antibody has been tested by flow cytometric analysis to detect a mouse IgA primary antibody and can be used at less than or equal to 0.2 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
Spezifikation
Spezifikation
| Antigen | Mouse IgA |
| Anwendungen | Flow Cytometry |
| Klassifikation | Monoclonal |
| Klon | 11-44-2 |
| Konzentration | 0.1 mg/mL |
| Konjugat | PE |
| Zusammensetzung | PBS with 0.09% sodium azide; pH 7.2 |
| Gen | Igha |
| Gen-Zugriffsnummer | P01878 |
| Gen-Alias | IgA; Igh-2; Igha; immunoglobulin heavy constant alpha; ImmunoglobulinA |
| Mehr anzeigen |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.