Learn More
IGF2BP3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Artikelnummer. 16863384 Alle ansehen Proteintech Produkte
Change view
Click to view available options
Menge:
150 μL
20 μL
Packungsgröße:
150 Mikroliter
20 Mikroliter
Artikelnummer. Menge unitSize
16863384 20 μL 20 Mikroliter
16853384 150 μL 150 Mikroliter
2 options
Artikelnummer. 16863384
Marke: Proteintech 146421AP20UL
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Rabbit Polyclonal Antibody
Both isoforms (64 kDa and 22 kDa) of IGFBP3 can be recognized by this antibody. IGFBP3 is nuclear and cytoplasmic in staining.
Insulin-like growth factor-II messenger RNA (mRNA)-binding protein-3 (IMP-3), also known as K homology domain-containing protein overexpressed in cancer (KOC) and L523S, is a member of the insulin-like growth factor-II mRNA-binding protein family and is expressed during embryogenesis and in some maligcies. IMP-3 is expressed in maligt melanoma but not in benign nevi, even when dysplastic features are present; IMP-3 is expressed in a significantly higher proportion of melanomas than Spitz nevi; and IMP-3 is expressed in metastatic melanomas significantly more than in thin melanomas. IMP-3 appears to be involved in the progression of maligt melanoma and may play an important role in the regulation of the biologic behavior of this tumor.
Spezifikation
Antigen IGF2BP3
Anwendungen Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin)
Klassifikation Polyclonal
Konzentration 0.15 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gen IGF2BP3
Gen-Zugriffsnummer O00425, Q9CPN8
Gen-Alias hKOC, IGF II mRNA binding protein 3, IGF2 mRNA binding protein 3, IGF2BP3, IMP 3, IMP3, KOC1, VICKZ family member 3, VICKZ3
Gensymbole IGF2BP3
Wirtsspezies Rabbit
Immunogen IGF2BP3 Fusion Protein Ag6226
Reinigungsverfahren Antigen Affinity Chromatography
Menge 20 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 10643, 140488
Zielspezies Mouse, Human
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Liquid
Isotype IgG