missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
IGFBP-rp1/IGFBP-7 Monoclonal specifically detects IGFBP-rp1/IGFBP-7 in Human samples. It is validated for Immunohistochemistry/Immunohistochemistry (Paraffin).
Specifications
Specifications
| Anwendungen | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Klon | 115 |
| Konjugat | PE |
| Zusammensetzung | PBS |
| Gen-Alias | angiomodulin, FSTL2, IBP-7, IGFBP-7PGI2-stimulating factor, IGFBP-7v, IGFBP-rP1, IGFBPRP1, insulin-like growth factor binding protein 7, insulin-like growth factor-binding protein 7, MAC25 protein, MAC25IGF-binding protein 7, Prostacyclin-stimulating factor, PSFAGM, TAF, Tumor-derived adhesion factor |
| Reinigungsverfahren | Protein A purified |
| Regulatorischer Status | RUO |
| Gen-ID (Entrez) | 3490 |
| Inhalt und Lagerung | Store at 4C in the dark. |
| Produkttyp | Antibody |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?