missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ IgM ELISA Development Kit
Alle ansehen Bio Techne Produkte
Click to view available options
Menge:
1 Kit (für 6 Platten)
Packungsgröße:
1 Set
Specifications
Specifications
| Assay-Bereich | 0.2 - 200 ng/mL |
| Produkttyp | ELISA Development Kit |
| Probentyp | Serum, Plasma, Cell Culture Supernatant |
| Zur Verwendung mit (Anwendung) | ELISA |
| Gen-ID (Entrez) | 3507 |
| Gensymbol | IGHM |
| Menge | 1 Kit (für 6 Platten) |
| Forschungsgebiet | B Cell Development and Differentiation Markers, Immunology |
| Lagerungsbedingungen | Storage of components varies. See protocol for specific instructions. |
| Synonym | AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction