Learn More
Invitrogen™ IL-1 beta Monoclonal Antibody (DF8)
GREENER_CHOICE
Artikelnummer. 11858721
Change view
Click to view available options
Menge:
500 μg
Packungsgröße:
500 Mikrogramm
Artikelnummer. Menge unitSize
11858721 500 μg 500 Mikrogramm
1 options
Artikelnummer. 11858721
Marke: Invitrogen™ MP425
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Mouse Monoclonal Antibody
MP425 targets IL-1 beta in WB applications and shows reactivity with Porcine samples. The MP425 immunogen is recombinant porcine IL-1 beta. MP425 detects IL-1 beta which has a predicted molecular weight of approximately 18 kDa. This product is a Low Endotoxin formulation. This product has been tested for endotoxins by limulus amoebocyte lysate (LAL) assay and contains an endotoxin concentration of less than or equal to 10 endotoxin units per milligram (EU/mg).
Interleukin-1 beta (IL-1 beta) is a proinflammatory cytokine expressed by monocytes, macrophages, and dendritic cells. IL-1 beta is synthesized in response to inflammatory stimuli as a 31 kDa inactive pro-form that accumulates in the cytosol. Cleavage of pro-IL-1 beta into the active 17 kDa protein requires the activation of inflammasomes, which are multi-protein complexes that respond to pathogens, stress conditions, and other danger signals. Inflammasome activation triggers the processing of the caspase-1 precursor into its active form, which in turn cleaves pro-IL-1 beta. IL-1 beta lacks a signal sequence peptide for classical ER/Golgi pathway and is secreted alongside caspase-1 via an alternate and incompletely understood mechanism. Although IL-1 beta is most often secreted in its active form, secretion of the uncleaved protein may be detectable under some biological conditions. IL-1 beta signals through two receptors, IL-1RI and IL-1RII, both of which are shared with IL-1 alpha. IL-1 beta activity can be moderated by IL-1 Receptor Antagonist (IL-1RA), a protein produced by many cell types that blocks receptor binding through competitive inhibition. IL-1 beta play an important role in innate host defense by triggering the production of other proinflammatory cytokines in target cells and initiating acute-phase responses to infection and injury. Elevated levels of IL-1 beta have been associated with many chronic inflammatory conditions IL-1 beta neutralizing antibodies potential therapeutic value.
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen IL-1 beta
Anwendungen Western Blot
Klassifikation Monoclonal
Klon DF8
Konzentration 1.0 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with no preservative
Gen IL1B
Gen-Zugriffsnummer P26889
Gen-Alias catabolin; cytokine; Hematopoietin 1 (H1); IFN beta inducing factor; il 1b; IL 1β; IL-1; IL1 B; IL-1 beta; IL-1 precursor; IL1B; IL-1B; IL1B1; IL1beta; IL-1beta; IL1-BETA; IL1F2; IL1β; ILN; Interleukin; interleukin 1 beta; Interleukin 1 beta precursor; interleukin 1, beta; interleukin 1, beta 1; interleukin 1-beta; interleukin*1*beta; Interleukin1 beta; interleukin-1 beta; interleukin-1 beta precursor; interleukin-1 beta precursor (AA -113 to 153); interleukin-1 beta proprotein; interleukin-1b; interleukin-1beta; interleukin-beta; LAF; lymphocyte proliferation-potentiating factor; Osteoclast activating factor (OAF); preinterleukin 1 beta; Pro interleukin 1 beta; prointerleukin-1 beta; pro-interleukin-1-beta
Gensymbole IL1B
Wirtsspezies Mouse
Immunogen Recombinant porcine IL-1 beta.
Reinigungsverfahren Protein G
Menge 500 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 397122
Zielspezies Pig
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Liquid
Isotype IgG1 κ