missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-1 RAcP/IL-1 R3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
168.00 €
Spezifikation
| Antigen | IL-1 RAcP/IL-1 R3 |
|---|---|
| Konzentration | 1 mg/mL |
| Verdünnung | Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 5 μg/mL |
| Anwendungen | Western Blot |
| Klassifikation | Polyclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18507730
|
Novus Biologicals
NBP1-76924-0.025mg |
0.025 mg |
168.00 €
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
IL-1 RAcP/IL-1 R3 Polyclonal specifically detects IL-1 RAcP/IL-1 R3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Spezifikation
| IL-1 RAcP/IL-1 R3 | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 5 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Cytokine Research, Immunology | |
| PBS with 0.02% Sodium Azide | |
| C3orf13IL-1RAcPIL1R3Interleukin-1 receptor 3, FLJ37788, IL-1 receptor accessory protein, IL-1R3, IL-1R-3, interleukin 1 receptor accessory protein, interleukin-1 receptor accessory protein, interleukin-1 receptor accessory protein beta | |
| IL1RAP | |
| IgG | |
| Affinity Purified | |
| It has no cross activity to other member in the IL-1 receptor family. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| AF029213 | |
| 3556 | |
| Antibody was raised against a peptide corresponding to 22 amino acids near the carboxy terminus of human IL-1RAcP. The immunogen is located within the last 50 amino acids of IL-1RAcP. Amino Acid Squence: KKSPRRSSSDEQGLSYSSLKNV | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| 63 kDa |
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel