Learn More
IL-10R alpha Antibody (RM0101-9E12), HRP, Novus Biologicals™
Hamster Monoclonal Antibody
Marke:  Novus Biologicals NBP1-22459H
Artikelnummer. 18217552
  • 625.00 € / 0.10 Milliliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
IL-10R alpha Monoclonal antibody specifically detects IL-10R alpha in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
IL-10R alpha
Monoclonal
HRP
PBS with No Preservative
IL10RA
Mouse recombinant protein of the Extracellular domain of IL-10R alpha
0.1 mL
Primary
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a hamster immunized with mouse recombinant protein of extracellular domain of IL-10R alpha.
Store at 4C in the dark.
IgG
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
RM0101-9E12
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
CD210 antigen, CDw210a, HIL-10R, IBD28, IL-10 receptor subunit alpha, IL-10R subunit 1, IL-10R1, IL-10RA, IL10RIL-10R subunit alpha, interleukin 10 receptor, alpha, interleukin-10 receptor alpha chain, Interleukin-10 receptor subunit 1, interleukin-10 receptor subunit alpha
Hamster
Protein A or G purified
Cytokine Research
3587
Mouse
Purified
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen