missing translation for 'onlineSavingsMsg'
Learn More

IL-12 Antibody (briakinumab) [Janelia Fluor™ 635], Novus Biologicals™

Artikelnummer. 30169684
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Menge:
0.1 mL
missing translation for 'unitSize'
0.10 Milliliter
Product Code. Menge unitSize
30169684 0.1 mL 0.10 Milliliter
1 missing translation for 'options'
This item is not returnable. View return policy

Product Code. 30169684

missing translation for 'mfr': Novus Biologicals (Bio-Techne) NBP328598JF635

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Human Monoclonal Antibody

IL-12 Monoclonal antibody specifically detects IL-12 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-12
Anwendungen Flow Cytometry, ELISA, Functional Assay
Klassifikation Monoclonal
Klon briakinumab
Konjugat Janelia Fluor 635
Zusammensetzung 50mM Sodium Borate
Gen-Alias CLMF p35, Cytotoxic lymphocyte maturation factor 35 kDa subunit, IL-12 subunit p35, IL-12, subunit p35, interleukin 12, p35, interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocytematuration factor 1, p35), interleukin-12 alpha chain, interleukin-12 subunit alpha, natural killer cell stimulatory factor 1, 35 kD subunit, NF cell stimulatory factor chain 1, NK cell stimulatory factor chain 1, NKSF1, p35
Wirtsspezies Human
Immunogen IL-12
Reinigungsverfahren Protein A purified
Menge 0.1 mL
Regulatorischer Status RUO
Forschungsgebiet Alzheimers Research, Immunology, Innate Immunity, Neurodegeneration
Primär oder sekundär Primary
Gen-ID (Entrez) 3592
Zielspezies Human
Inhalt und Lagerung Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.