missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-12 p40 Recombinant Rabbit Monoclonal Antibody (240)

Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA560511
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
IL-12 p40 Recombinant Monoclonal Antibody for ELISA
IL12B (IL-12 p40) encodes a subunit of interleukin 12, a cytokine that acts on T and natural killer cells, and has a broad array of biological activities. Interleukin 12 is a disulfide-linked heterodimer composed of the 40 kDa cytokine receptor like subunit encoded by this gene, and a 35 kDa subunit encoded by IL12A. This cytokine is expressed by activated macrophages that serve as an essential inducer of Th1 cells development. This cytokine has been found to be important for sustaining a sufficient number of memory/effector Th1 cells to mediate long-term protection to an intracellular pathogen. Overexpression of this gene was observed in the central nervous system of patients with multiple sclerosis (MS), suggesting a role of this cytokine in the pathogenesis of the disease. The promoter polymorphism of this gene has been reported to be associated with the severity of atopic and non-atopic asthma in children.
Spezifikation
| IL-12 p40 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P43432 | |
| Il12b | |
| Recombinant Mouse IL-12 p40/IL12B protein (Met1-Ser335). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| ELISA | |
| 240 | |
| Unconjugated | |
| Il12b | |
| CLMF; CLMF p35; CLMF p40; CLMF2; cytotoxic lymphocyte maturation factor 40 kDa subunit; Il12; IL12 p40; IL-12 p40 subunit; IL-12 subunit p35; IL-12 subunit p40; IL12, subunit p40; IL-12.p40; IL12A; IL-12A; Il12b; IL-12B; Il12p40; IL-12p40; IL-12p40 subunit; IMD28; IMD29; Interleukin; interleukin 12 p40 subunit; interleukin 12 subunit p40; interleukin 12, p40; interleukin 12B; interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocyte maturation factor 2, p40); interleukin-12 beta chain; interleukin-12 beta chain precursor; interleukin-12 p40; interleukin-12 p40 chain; interleukin-12 p40 subunit; interleukin-12 subunit beta; interleukin-12 subunit p40; interleukin-12p40; interleukin-23 p40 subunit; natural killer cell stimulatory factor, 40 kD subunit; NK cell stimulatory factor chain 2; NKSF; NKSF1; NKSF2; p40; RP23-388G23.1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 16160 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur