missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-13 Monoclonal Antibody (eBio13A), PE-Cyanine7, eBioscience™, Invitrogen™

Artikelnummer. p-7090486
Change view
Click to view available options
Menge:
100 μg
25 μg
Packungsgröße:
100 Mikrogramm
25 Mikrogramm
Artikelnummer. Menge unitSize
15528636 25 μg 25 Mikrogramm
15538636 100 μg 100 Mikrogramm
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 15528636

Marke: Invitrogen™ 25713380

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rat Monoclonal Antibody

Description: The eBio13A antibody reacts with mouse IL-13. IL-13 is a cytokine produced mainly by Th2 cells, but also by antigen-primed CD8 T cells. IL-13 has a strong involvement in allergic inflammation and parasitic clearing and in cancer models has been shown to have either inhibitory or stimulatory activity depending on the tumor. In humans, IL-13 is found to play a role in isotype switching in B cells. IL-13 is implicating in down modulating macrophage activity, through the reduction of pro-inflammatory cytokines (IL-1, IL-6, IL-8, IL-10, IL-12) Applications Reported: This eBio13A antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This eBio13A antibody has been tested by intracellular staining of stimulated mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat.

IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen IL-13
Anwendungen Flow Cytometry
Klassifikation Monoclonal
Klon eBio13A
Konzentration 0.2 mg/mL
Konjugat PE-Cyanine7
Zusammensetzung PBS with 0.09% sodium azide; pH 7.2
Gen Il13
Gen-Zugriffsnummer P20109
Gen-Alias ALRH; ALRH antibody; BHR1; il 13; Il13; Il-13; IL-13; cytokine; ILN; Interleukin; interleukin 13; Interleukin13; interleukin-13; interleukin-13 precursor; L13; MGC116786; MGC116788; MGC116789; NC30; NC30 antibody; P600; P600 antibody; T-cell activation protein P600
Gensymbole Il13
Wirtsspezies Rat
Reinigungsverfahren Affinity chromatography
Menge 25 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 16163
Zielspezies Mouse
Inhalt und Lagerung 4°C, store in dark, DO NOT FREEZE!
Produkttyp Antibody
Form Liquid
Isotype IgG1 κ
Mehr anzeigen Weniger anzeigen

For Research Use Only.

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.