Learn More
IL-17RA/IL-17R Antibody (CL10309) [DyLight 405], Novus Biologicals™
Alle ansehen Bio Techne ProdukteBeschreibung
IL-17RA/IL-17R Monoclonal antibody specifically detects IL-17RA/IL-17R in Human samples. It is validated for Western Blot
Spezifikation
Spezifikation
| Antigen | IL-17RA/IL-17R |
| Anwendungen | Western Blot |
| Klassifikation | Monoclonal |
| Klon | CL10309 |
| Konjugat | DyLight 405 |
| Zusammensetzung | 50mM Sodium Borate |
| Gen-Alias | CD217, CD217 antigen, CDw217interleukin 17 receptor, hIL-17R, IL-17 receptor A, IL-17RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A |
| Wirtsspezies | Mouse |
| Immunogen | This antibody was developed using a synthetic peptide derived from Q96F46, with the exact immunogen sequence remaining proprietary. |
| Reinigungsverfahren | Protein A purified |
| Mehr anzeigen |