Learn More
IL-18 R beta/IL-1 R7/ACPL Antibody (122), mFluor Violet 610 SE, Novus Biologicals™
Alle ansehen Bio Techne ProdukteBeschreibung
IL-18 R beta/IL-1 R7/ACPL Monoclonal antibody specifically detects IL-18 R beta/IL-1 R7/ACPL in Human samples. It is validated for ELISA, ELISA
Spezifikation
Spezifikation
| Antigen | IL-18 R beta/IL-1 R7/ACPL |
| Anwendungen | ELISA, Sandwich ELISA |
| Klassifikation | Monoclonal |
| Klon | 122 |
| Konjugat | mFluor Violet 610 SE |
| Verdünnung | ELISA, Sandwich ELISA Detection |
| Zusammensetzung | 50mM Sodium Borate |
| Gen-Alias | Accessory protein-like, ACPL, CD218 antigen-like family member B, CD218b, CD218b antigen, CDw218b, IL-18 receptor accessory protein, IL-18 receptor beta, IL-18RAcP, IL18RB, IL-18Rbeta, IL-18R-beta, IL-1R accessory protein-like, IL1R7, IL-1R7, IL-1R-7, IL-1RAcPL, interleukin 18 receptor accessory protein, Interleukin-1 receptor 7, interleukin-18 receptor accessory protein, Interleukin-18 receptor accessory protein-like, Interleukin-18 receptor beta, MGC120589, MGC120590 |
| Wirtsspezies | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-18 R beta/IL-1 R7/ACPL (Accession#: NP_003844.1; Met1-Gly357). |
| Mehr anzeigen |