Learn More
Invitrogen™ Human IL-29 (IFN lambda 1) Uncoated ELISA Kit
Beschreibung
The Human Interleukin 29 (IFN lambda 1) Uncoated ELISA Kit contains pre-matched antibody pairs, and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human IL-29. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Spezifikation
Spezifikation
| Zugriffsnummer | Gehäuse Q8IU54 |
| Assay-Bereich | 8 bis 1,000 pg/ml |
| Assay-Empfindlichkeit: | 8 pg/mL |
| Inhalt und Lagerung | Capture Antibody: Pre-titrated, purified antibody Detection Antibody: Pre-titrated, biotin-conjugated antibody Standard: Recombinant cytokine for generating standard curve and calibrating samples ELISA/ELISPOT Coating Buffer Powder: This uncoated ELISA kit may contain ELISA/ELISPOT Coating Buffer Powder (Reconstitute to 1L with dH20 and filter (0.22 μM)) or 10X PBS ELISA Coating Buffer (Dilute 1 part 10X Buffer into 9 parts dH20). Assay Diluent: 5X concentrated Detection enzyme: Pre-titrated Avidin-HRP Substrate Solution: Tetramethylbenzidine (TMB) Substrate Solution Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards 96 Well Plate: Corning Costar 9018 (included with product Cat. Nos. ending in suffixes -22, -44, -76, -86) |
| Konjugat | Biotin |
| Immunassay-Kit-Format | Uncoated ELISA Kit |
| Produkttyp | ELISA-Kit |
| Probentyp | Plasma, Serum, Überstand |
| Zielspezies | Human |
| Zur Verwendung mit (Anwendung) | ELISA |
| Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.