missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-3R alpha/CD123 Antibody (6H6), Alexa Fluor™ 647, Novus Biologicals™
Alle ansehen Bio Techne ProdukteDescription
IL-3R alpha/CD123 Monoclonal specifically detects IL-3R alpha/CD123 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Flow (Intracellular).
Specifications
Specifications
| Antigen | IL-3R alpha/CD123 |
| Anwendungen | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) |
| Klassifikation | Monoclonal |
| Klon | 6H6 |
| Konjugat | Alexa Fluor 647 |
| Verdünnung | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Flow (Intracellular) |
| Gen-Alias | CD123, CD123 antigen, hIL3Ra, hIL-3Ra, IL-3 receptor subunit alpha, IL3R, IL-3R subunit alpha, IL-3RA, IL-3R-alpha, IL3RAY, IL3RX, IL3RY, interleukin 3 receptor, alpha (low affinity), interleukin-3 receptor subunit alpha, MGC34174 |
| Gensymbole | IL3RA |
| Wirtsspezies | Mouse |
| Immunogen | Tissue cell preparation. IL3R transfected COS cells. |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?