missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-4 Antibody (BVD4-1D11), PerCP, Novus Biologicals™
Rat Monoclonal Antibody
Marke: Novus Biologicals NB100-64798PCP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
IL-4 Monoclonal specifically detects IL-4 in Mouse samples. It is validated for Flow Cytometry.
Spezifikation
| IL-4 | |
| Monoclonal | |
| PerCP | |
| B cell growth factor 1, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL-4B_cell stimulatory factor 1, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This is specific for mouse Interleukin-4 (IL-4). Interleukin-4 is a potent lymphoid growth factor, which stimulates the growth and survival of certain B and T cells. Clone BVD4-1D11 is reported to neutralise the effects of mouse IL-4 (3). | |
| Store at 4C in the dark. | |
| IgG2b |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| BVD4-1D11 | |
| Flow Cytometry | |
| IL4 | |
| Recombinant Mouse IL-4 | |
| 0.25 mL | |
| Adaptive Immunity, Asthma, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity | |
| 3565 | |
| Mouse | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur