Learn More
PeproTech IL-4 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Beschreibung
AA Sequence of recombinant protein: MHGCNDSPLR EIINTLNQVT EKGTPCTEMF VPDVLTATRN TTENELICRA SRVLRKFYFP RDVPPCLKNK SGVLGELRKL CRGVSGLNSL RSCTVNESTL TTLKDFLESL KSILRGKYLQ SCTSMS. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Rat IL-4. Anti-Rat IL-4-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Rat IL-4 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Rat IL-4 (500-P94) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Rat IL-4. Western Blot: To detect Rat IL-4 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Rat IL-4 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P94BT-1MG will be provided as 2 x 500 μg
Spezifikation
Spezifikation
| Antigen | IL-4 |
| Anwendungen | ELISA, Western Blot |
| Klassifikation | Polyclonal |
| Konzentration | 0.1-1.0 mg/mL |
| Konjugat | Biotin |
| Zusammensetzung | PBS with no preservative |
| Gen | Il4 |
| Gen-Zugriffsnummer | P20096 |
| Gen-Alias | B cell growth factor 1; B_cell stimulatory factor 1; B-cell growth factor 1; B-cell IgG differentiation factor; B-cell stimulatory factor 1; BCGF1; BCGF-1; binetrakin; BSF1; BSF-1; cytokine; H-IL-4; IGG1 induction factor; IL2; IL4; Il-4; Il4e12; ILN; Interleukin; interleukin 4; interleukin 4 variant 2; interleukin 4 variant IL-4delta2; interleukin 4 variant IL-4delta3; interleukin 4 variant IL-4int2A; interleukin 4 variant IL-4int3A; interleukin 4 variant IL-4int3B; Interleukin4; interleukin-4; interleukin-4 precursor; lymphocyte stimulatory factor 1; MGC79402; M-IL-4; Pitrakinra |
| Gensymbole | Il4 |
| Mehr anzeigen |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.