missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-4I1 Antibody (CL9071) [Alexa Fluor™ 405], Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP2-88929AF405
This item is not returnable.
View return policy
Description
IL-4I1 Monoclonal antibody specifically detects IL-4I1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| IL-4I1 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| FIG1, Fig-1 protein, hFIG1, IL4-induced protein 1, interleukin 4 induced 1, interleukin four induced 1, LAAO, L-amino-acid oxidase, LAO, UNQ636/PRO1265 | |
| This antibody was developed using a synthetic peptide derived from Q96RQ9, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Apoptosis, Cell Biology | |
| 259307 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL9071 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction