missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-5 Monoclonal Antibody (39D10), Biotin
GREENER_CHOICE

Artikelnummer. 11874281
Change view
Click to view available options
Menge:
250 μg
Packungsgröße:
250 Mikrogramm
Artikelnummer. Menge unitSize
11874281 250 μg 250 Mikrogramm
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 11874281

Marke: Invitrogen™ M550B

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rat Monoclonal Antibody

M550B targets IL-5 in ELISA, and WB applications and shows reactivity with Human samples. The M550B immunogen is recombinant human IL-5. M550B detects IL-5 which has a predicted molecular weight of approximately 13 kDa. This antibody is produced by injecting Rat IgG secreting hybridoma cells into the peritoneum of mice. The resulting ascites is collected from the mice and the antibody is purified. The M550B IL1b antibody (biotinylated conjugate of clone 39D10) has successfully been paired as the detection antibody in a sandwich ELISA with capture antibody M551 (clone 5A10). Typical dilutions for sandwich ELISA include 1 μg/mL for coating and 0.125 - 0.25 μg/mL for detection. Biotinylated antibody M550B (clone 39D10) and antibody M551 (clone 5A10) have successfully been used in combination with recombinant IL5 protein SIL5 in ELISA applications.

IL-5 is a cytokine that acts as a growth and differentiation factor for both B cells and eosinophils. IL-5 cytokine is a main regulator of eosinopoiesis, eosinophil maturation and activation. The elevated production of this cytokine is reported to be related to asthma or hypereosinophilic syndromes. The receptor of IL-5 cytokine is a heterodimer, whose beta subunit is shared with the receptors for interleukine 3 (IL3) and colony stimulating factor 2 (CSF2/GM-CSF). IL-5, together with those for interleukin 4 (IL4), interleukin 13 (IL13), and CSF2, form a cytokine gene cluster on chromosome 5. IL-5, IL4, and IL13 are found to be regulated coordinately by long-range regulatory elements spread over 120 kilobases on chromosome 5q31.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen IL-5
Anwendungen ELISA, Western Blot
Klassifikation Monoclonal
Klon 39D10
Konzentration 0.5 mg/mL
Konjugat Biotin
Zusammensetzung PBS with 4% BSA and proprietary Preservative
Gen Il5
Gen-Zugriffsnummer P05113
Gen-Alias B-cell differentiation factor I; B-cell growth factor II; BCGF-II; colony-stimulating factor, eosinophil; cytotoxic T-lymphocyte inducer; EDF; Eosinophil differentiation factor; IL5; IL-5; ILN; Interleukin; Interleukin 5; Interleukin 5 (colony-stimulating factor eosinophil); Interleukin 5 (colony-stimulating factor, eosinophil); Interleukin5; interleukin-5; T-cell replacing factor; TRF
Gensymbole Il5
Wirtsspezies Rat
Immunogen Recombinant human IL-5.
Reinigungsverfahren Caprylic acid precipitation
Menge 250 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 3567
Zielspezies Human
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Liquid
Isotype IgG2a
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.