missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ILKAP Polyclonal Antibody
Sheep Polyclonal Antibody
Marke: Invitrogen™ PA5148868
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute at 0.2 mg/mL in sterile PBS.
The protein encoded by this gene is a protein serine/threonine phosphatase of the PP2C family. This protein can interact with integrin-linked kinase (ILK/ILK1), a regulator of integrin mediated signaling, and regulate the kinase activity of ILK. Through the interaction with ILK, this protein may selectively affect the signaling process of ILK-mediated glycogen synthase kinase 3 beta (GSK3beta), and thus participate in Wnt signaling pathway.
Spezifikation
| ILKAP | |
| Polyclonal | |
| Unconjugated | |
| ILKAP | |
| 0710007A14Rik; 1600009O09Rik; EC 3.1.3.16; ILK associated serine/threonine phosphatase; Ilkap; ILKAP2; ILKAP3; integrin-linked kinase associated phosphatase; integrin-linked kinase-associated serine/threonine phosphatase; integrin-linked kinase-associated serine/threonine phosphatase 2C; PP2Cdelta; PP2C-DELTA; PPM1O; protein phosphatase 2C; protein phosphatase 2c, delta isozyme; protein phosphatase, Mg2+/Mn2+ dependent 1O | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 64538, 67444, 80895 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q8R0F6, Q9H0C8, Q9Z1Z6 | |
| ILKAP | |
| E. coli-derived recombinant human ILKAP, Met1-His392. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur