missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ILT7/CD85g/LILRA4 Antibody (656688) [Janelia Fluor™ 549], Novus Biologicals™
Alle ansehen Bio Techne ProdukteBeschreibung
ILT7/CD85g/LILRA4 Monoclonal antibody specifically detects ILT7/CD85g/LILRA4 in Human samples. It is validated for Flow Cytometry, CyTOF
Spezifikation
Spezifikation
| Antigen | ILT7/CD85g/LILRA4 |
| Anwendungen | Flow Cytometry, CyTOF |
| Klassifikation | Monoclonal |
| Klon | 656688 |
| Konjugat | Janelia Fluor 549 |
| Verdünnung | Flow Cytometry, CyTOF-ready |
| Zusammensetzung | 50mM Sodium Borate |
| Gen-Alias | CD85 antigen-like family member G, CD85g, CD85g antigen, ILT-7, ILT7MGC129598, Immunoglobulin-like transcript 7, leukocyte immunoglobulin-like receptor subfamily A member 4, leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 4, leukocyte immunoglobulin-like receptor, subfamily A (without TM domain), member4, MGC129597 |
| Wirtsspezies | Mouse |
| Immunogen | Mouse myeloma cell line NS0-derived recombinant human LILRA4/CD85g/ILT7, Glu24-Asn446, Accession # P59901 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?