Learn More
Influenza A H1N1 Hemagglutinin Antibody (10), Alexa Fluor™ 700, Novus Biologicals™
Rabbit Monoclonal Antibody
Marke: Novus Biologicals NBP3-06578AF700
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Influenza A H1N1 Hemagglutinin Monoclonal antibody specifically detects Influenza A H1N1 Hemagglutinin in Influenza virus N1 samples. It is validated for Western Blot
Spezifikation
| Influenza A H1N1 Hemagglutinin | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Influenza H1N1 (A/California/04/2009) Hemagglutinin. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| 10 | |
| Western Blot | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Influenza virus N1 | |
| Purified |