missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Influenza A H3N2 HA (A/TW/3446/02) Recombinant Rabbit Monoclonal Antibody (HL2393)

Beschreibung
Influenza A H3N2 HA (A/TW/3446/02) Recombinant Monoclonal Antibody for Western Blot, ICC/IF
Influenza viruses consists of three types (Influenza A, B, and C), which make up three of the five genera in the Orthomyxoviridae family. Commonly know as the flu, influenza strains can affect both birds and mammals. Influenza strains form roughly spherical particles and are similar in composition with viral envelope containing two main types of glycoproteins (hemagglutinin and neuraminidase), wrapped around the central core of the viral RNA genome and other viral proteins that package and protect the RNA. Unusually for a virus, the influenzavirus genome is not a single piece of nucleic acid; rather a seven to eight segmented fragments of negative-sense RNA with each fragment expressing different viral proteins. Influenza spreads seasonally, resulting in about three to five million yearly cases of severe illness and about 250,000 to 500,000 deaths annually or into the millions in pandemic years.
Spezifikation
Spezifikation
| Antigen | Influenza A H3N2 HA (A/TW/3446/02) |
| Anwendungen | Western Blot, Immunocytochemistry |
| Klassifikation | Recombinant Monoclonal |
| Klon | HL2393 |
| Konzentration | 1 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with no preservative |
| Gen-Alias | flu hemagglutinin; H3N2 HA; H3N2 hemagglutinin; Influenza H3N2; influenza hemagglutinin |
| Wirtsspezies | Rabbit |
| Immunogen | Recombinant fragment of Influenza A virus H3N2 HA (Hemagglutinin) (Influenza A virus (A/TW/3446/02(H3N2)). |
| Mehr anzeigen |
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?