Learn More
InSphero Incubox™
Beschreibung
The Incubox™ is a specially engineered container which creates a high-humidity microclimate inside conventional cell culture incubators and protects your sensitive plates from large humidity drops and fluctuations – the major cause for evaporation effects.
- Maintain a stable medium volume and accurate compound concentration due to minimal evaporation over time.
- Reduce well-to-well variation by minimizing evaporation-induced reagent gradients across the plate.
- Use all wells of your plate by eliminating edge effects in your culture plate.
- Save on plasticware by using up to 38% more wells of your plate.
Spezifikation
Spezifikation
| Zur Verwendung mit (Geräte) | Inkubatoren |
| Material | Glas |
| Typ | Incubation Microplate Container |