missing translation for 'onlineSavingsMsg'
Learn More

Insulin Antibody (IRDN/1980R), Allophycocyanin, Novus Biologicals™

Rabbit Monoclonal Antibody

Marke:  Novus Biologicals NBP3-08836APC

Product Code. 18647087

  • 713.00 € / 0.10 Milliliter

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Description

Description

Insulin Monoclonal antibody specifically detects Insulin in Human, Mouse, Porcine, Bovine, Rabbit samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Insulin
Monoclonal
APC
IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
Recombinant human full-length Insulin protein (IRDN) (Uniprot: P01308)
0.1 mL
Diabetes Research, Immune System Diseases, Immunology, Lipid and Metabolism, Stem Cell Markers
3630
Store at 4°C in the dark.
IgG
Immunohistochemistry (Paraffin)
IRDN/1980R
PBS
Rabbit
Protein A or G purified
RUO
Primary
Human, Mouse, Pig, Bovine, Rabbit
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.