Learn More
Insulin R/CD220 Antibody (83-14), Alexa Fluor™ 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Marke:  Novus Biologicals NBP2-81038AF488
Artikelnummer. 18300540
  • 587.00 € / 0.10 Milliliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
Insulin R/CD220 Monoclonal specifically detects Insulin R/CD220 in Human, Porcine, Bovine, Sheep samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Insulin R/CD220
Monoclonal
Alexa Fluor 488
50mM Sodium Borate
INSR
This antibody was raised by immunizing mice with IM-9 lymphocytes followed by purified insulin receptor.
0.1 mL
Diabetes Research, Lipid and Metabolism, Neuroscience, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases
3643
Human, Pig, Bovine, Sheep
Purified
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
83-14
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
CD 220, CD220, CD220 antigen, EC 2.7.10, EC 2.7.10.1, HHF5, insulin receptor, IR
Rabbit
Protein A purified
RUO
Primary
Recognises the extracellular domain of the Insulin R/CD220. Amino acids 469 - 592 (exon 7 and 8) of Insulin R/CD220.
Store at 4°C in the dark.
IgG κ
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen