missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha V beta 3 Antibody (27.1 (VNR-1)), Alexa Fluor™ 488, Novus Biologicals™
Alle ansehen Bio Techne ProdukteDescription
Integrin alpha V beta 3 Monoclonal antibody specifically detects Integrin alpha V beta 3 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
Specifications
| Antigen | Integrin alpha V beta 3 |
| Anwendungen | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Klassifikation | Monoclonal |
| Klon | 27.1 (VNR-1) |
| Konjugat | Alexa Fluor 488 |
| Verdünnung | Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen |
| Zusammensetzung | 50mM Sodium Borate |
| Gen-Alias | antigen identified by monoclonal L230, CD51, CD51 antigen, integrin alpha-V, integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51), MSK8, Vitronectin receptor subunit alpha, VNRADKFZp686A08142 |
| Wirtsspezies | Mouse |
| Immunogen | This antibody is specific to Integrin Alpha VBeta 3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?