Learn More
Integrin beta 2/CD18 Antibody (60.3), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP3-12030PCP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Integrin beta 2/CD18 Monoclonal antibody specifically detects Integrin beta 2/CD18 in Human samples. It is validated for Western Blot, ELISA, Functional Assay (Inhibition), Neutralization
Spezifikation
| Neutralization | |
| Mouse | |
| Primary |
| Western Blot, ELISA, Functional (Inhibition), Block/Neutralize | |
| Apoptosis, Cytokine Research, Immunology, Phospho Specific |