Learn More
Integrin beta 4/CD104 Antibody (CL6972) [PerCP], Novus Biologicals™
Mouse Monoclonal Antibody
Marke:  Novus Biologicals NBP2-76524PCP
Artikelnummer. 30307631
  • 836.00 € / 0.10 Milliliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
Integrin beta 4/CD104 Monoclonal antibody specifically detects Integrin beta 4/CD104 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Spezifikation
Specifications
Integrin beta 4/CD104
Monoclonal
PerCP
CD104, CD104 antigen, GP150, integrin beta-4, integrin beta-4 subunit, integrin, beta 4
This antibody was developed using a recombinant protein derived from P16144, with the exact immunogen sequence remaining proprietary.
0.1 mL
Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction
3691
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL6972
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified