missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
Integrin beta 4/CD104 Monoclonal antibody specifically detects Integrin beta 4/CD104 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Spezifikation
Spezifikation
| Antigen | Integrin beta 4/CD104 |
| Anwendungen | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Klassifikation | Monoclonal |
| Klon | CL6973 |
| Konjugat | FITC |
| Zusammensetzung | PBS |
| Gen-Alias | CD104, CD104 antigen, GP150, integrin beta-4, integrin beta-4 subunit, integrin, beta 4 |
| Wirtsspezies | Mouse |
| Immunogen | This antibody was developed using a recombinant protein derived from P16144, with the exact immunogen sequence remaining proprietary. |
| Reinigungsverfahren | Protein A purified |
| Mehr anzeigen |
Name des Produkts
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?