missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IP3 Competitive ELISA Kit
Click to view available options
:
96 Tests
Beschreibung
Inositol 1,4,5-trisphosphate (IP3) is a crucial intracellular signaling molecule that plays an essential role in various cellular processes by regulating calcium release from the endoplasmic reticulum. IP3 is generated through the hydrolysis of phosphatidylinositol 4,5-bisphosphate (PIP2) by the enzyme phospholipase C (PLC) following receptor activation. The released IP3 binds to its receptors, inositol 1,4,5-trisphosphate receptors (ITPRs), which are located on the endoplasmic reticulum membrane. This binding triggers the release of Ca2+ ions into the cytoplasm, leading to numerous downstream effects, including muscle contraction, secretion, metabolism, and cell growth. ITPRs are encoded by different genes leading to multiple isoforms (ITPR1, ITPR2, and ITPR3), each showing distinct tissue distribution and regulatory properties. Understanding the specific functions and regulatory mechanisms of IP3 and ITPRs is critical for elucidating their roles in health and disease.
Spezifikation
Spezifikation
| Assay-Bereich | 15.63 to 1000 pg/mL |
| Assay-Empfindlichkeit: | 9.38 pg/mL |
| Konjugat | HRP |
| Produkttyp | ELISA |
| Probentyp | Plasma, Serum |
| Zur Verwendung mit (Geräte) | Colorimetric Microplate Reader |
| Interassay-VK | <10 |
| Intraassay-VK | <10 |
| Kitinhalt | Micro ELISA Plate (Dismountable), Reference Standard, Concentrated Biotinylated Detection Ab(100x), Concentrated HRP Conjugate (100x), Reference Standard & Sample Diluent, Biotinylated Detection Ab Diluent, HRP Conjugate Diluent, Concentrated Wash Buffer(25x), Substrate Reagent, Stop Solution, Plate Sealer, Product Description, Certificate of Analysis |
| Marker oder Farbstoff | Biotin |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur