missing translation for 'onlineSavingsMsg'
Learn More

IP6K1 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Artikelnummer. 18500619 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
0.1 mL
Unit Size:
0.10 Milliliter
Product Code. Menge unitSize
18500619 0.1 mL 0.10 Milliliter
1 options
This item is not returnable. View return policy

Product Code. 18500619

Brand: Novus Biologicals NBP318438AF488

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IP6K1 Polyclonal antibody specifically detects IP6K1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen IP6K1
Anwendungen Western Blot
Klassifikation Polyclonal
Konjugat Alexa Fluor 488
Verdünnung Western Blot
Zusammensetzung 50mM Sodium Borate
Gen-Alias ATP:1D-myo-inositol-hexakisphosphate phosphotransferase, EC 2.7.4.21, inositol hexakisphosphate kinase 1, Inositol hexaphosphate kinase 1KIAA0263IHPK1PiUS, InsP6 kinase 1, MGC9925, Pi uptake stimulator
Wirtsspezies Rabbit
Immunogen Recognizes region between residues Between Between 75 and 125..
Reinigungsverfahren Affinity purified
Menge 0.1 mL
Regulatorischer Status RUO
Forschungsgebiet Neuroscience, Neurotransmission, Signal Transduction
Primär oder sekundär Primary
Gen-ID (Entrez) 9807
Zielspezies Human
Inhalt und Lagerung Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.