Learn More
Isocitrate Dehydrogenase 1/IDH1 Antibody (IDH1.R132H/7277R), Alexa Fluor™ 532, Novus Biologicals™
Alle ansehen Bio Techne ProdukteBeschreibung
Isocitrate Dehydrogenase 1/IDH1 Monoclonal antibody specifically detects Isocitrate Dehydrogenase 1/IDH1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Spezifikation
Spezifikation
| Antigen | Isocitrate Dehydrogenase 1/IDH1 |
| Anwendungen | Immunohistochemistry (Paraffin) |
| Klassifikation | Monoclonal |
| Klon | IDH1.R132H/7277R |
| Konjugat | Alexa Fluor 532 |
| Zusammensetzung | 50 mM Sodium Borate |
| Gen-Alias | Cytosolic NADP-isocitrate dehydrogenase, EC 1.1.1.42, IDCD, IDH, IDP, IDPC, isocitrate dehydrogenase [NADP] cytoplasmic, isocitrate dehydrogenase 1 (NADP+), soluble, NADP(+)-specific ICDH, NADP-dependent isocitrate dehydrogenase, cytosolic, NADP-dependent isocitrate dehydrogenase, peroxisomal, Oxalosuccinate decarboxylase, PICD |
| Wirtsspezies | Rabbit |
| Immunogen | Recombinant fragment of human Isocitrate Dehydrogenase 1/IDH1 (exact sequence is proprietary) |
| Reinigungsverfahren | Protein A or G purified |
| Mehr anzeigen |