Learn More
Isopropyl Alcohol USP, Macron Fine Chemicals™
Artikelnummer. 15558074
Ansicht ändern
Click to view available options
Glas; 4 l
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer.
15558074 Glas; 4 l
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 15558074 Lieferant Avantor Macron Lieferanten-Nr. 303108
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen
Spezifikation
Chemischer Name oder Material Isopropyl Alcohol
CAS 67-63-0
Menge 4 L
Verpackung Glas
Summenformel C3H8O
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
InChI-Schlüssel KFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILES CC(C)O
IUPAC-Name Propan-2-ol
Molekulargewicht (g/mol) 60.096
PubChem CID 3776
ChEBI CHEBI:17824
Formelmasse 60.1
Güte BP/EP/FCC/JP/NF/USP