Beschreibung
Component of the cornified envelope of keratinocytes. May be involved in the interplay between adherens junctions and desmosomes. The function in the nucleus is not known.
Spezifikation
Spezifikation
| Antigen | kazrin |
| Anwendungen | Immunohistochemistry (Paraffin), Western Blot |
| Klassifikation | Polyclonal |
| Konzentration | 0.2 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gen | KAZN |
| Gen-Zugriffsnummer | Q5FWS6, Q674X7, Q69ZS8 |
| Gen-Alias | KAZ, kazrin, KIAA1026, RP1 21O18.1, RP1-21O18.1 |
| Gensymbole | Kazn |
| Mehr anzeigen |