missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KIR2DL1 Recombinant Human Monoclonal Antibody (2F3)

Human Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA560773
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
KIR2DL1 Recombinant Monoclonal Antibody for IHC (P), ELISA
This gene encodes a myotubularin-related protein. The encoded protein is a phosphoinositide phosphatase that specifically dephosphorylates phosphatidylinositol 3,5-biphosphate and phosphatidylinositol 3-phosphate. Mutations in this gene are correlated with autosomal dominant centronuclear myopathy. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 18.
Spezifikation
| KIR2DL1 | |
| Recombinant Monoclonal | |
| 1.91 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P43626 | |
| KIR2DL1 | |
| Recombinant Human KIR2DL1 protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG4SP |
| ELISA, Immunohistochemistry (Paraffin) | |
| 2F3 | |
| Unconjugated | |
| KIR2DL1 | |
| CD158 antigen-like family member A; CD158a; killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 1; killer cell immunoglobulin-like receptor 2DL1; killer cell immunoglobulin-like receptor two domains short cytoplasmic tail 1; killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 1; killer Ig receptor; killer inhibitory receptor 2-2-1; KIR221; KIR2DL1; KIR2DS1; KIR-K64; MHC class I NK cell receptor; natural killer cell receptor; Natural killer-associated transcript 1; NKAT; NKAT1; NKAT-1; p58 killer cell inhibitory receptor KIR-K64; p58 natural killer cell receptor clones CL-42/47.11; p58 NK cell inhibitory receptor NKR-K6; p58 NK receptor CL-42/47.11; p58.1; p58.1 MHC class-I-specific NK receptor | |
| Human | |
| Affinity chromatography | |
| RUO | |
| 3802 | |
| -20°C or -80°C if preferred | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur