Learn More
DiscoverX Kisspeptin-10/Metastin
Click to view available options
:
1 mg
Beschreibung
Kisspeptin-10 is a protein and potent endogenous agonist for the kisspeptin receptor, GPR54 (Ki=2.33nM). Kisspeptin-GPR54 signaling assists in gonadotropin-releasing hormone secretion at puberty. Kisspeptin-10 is the smallest fully bioactive fragment of kisspeptin, and was originally identified as a metastasis suppressor for melanoma and breast cancer. Provided as 1mg of dry powder.
Spezifikation
Spezifikation
| Menge | 1 mg |
| Produkttyp | Control Ligands |