missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Klebsiella aerogenes K15 Monoclonal Antibody (ZX3.1)

Mouse Monoclonal Antibody
Marke: Invitrogen™ MA518038
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This antibody reacts with Klebsiella aerogenes serotype 15.
Klebsiella are non-motile, rod-shaped, aerobic bacteria that possess a prominent polysaccharide capsule. Some of these bacteria produce an extracellular toxic complex that has been shown to be 'lethal for and produce extensive lung pathology in mice'. It is composed of 63% capsular polysaccharide, 30% lipopolysaccharide, and 7% protein; when introduced to experimental animals in sublethal doses, the animals built up immunization due to antibody production. Klebsiella pneumoniae is one of the most common Gram-negative bacteria seen by physicians worldwide. Pneumonias that are caused by Klebsiella pneumoniae are difficult to control; mortality rates have even been reported as up to 50% after antibiotic treatment. Nitrogen metabolism in Klebsiella aerogenes has also been studied throroughly.
Spezifikation
| Klebsiella aerogenes K15 | |
| Monoclonal | |
| 0.4 mg/mL | |
| PBS with 10 mM sodium azide | |
| Mouse | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG3 |
| ELISA | |
| ZX3.1 | |
| Unconjugated | |
| k aerogenes; K. aerogenes | |
| LPS membrane extract from K.aerogenes. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur