Beschreibung
KLF4 Monoclonal antibody specifically detects KLF4 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Spezifikation
Spezifikation
| Antigen | KLF4 |
| Anwendungen | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Klassifikation | Monoclonal |
| Klon | CL5785 |
| Konjugat | PE |
| Zusammensetzung | PBS |
| Gen-Alias | Epithelial zinc finger protein EZF, EZFendothelial Kruppel-like zinc finger protein, GKLFKrueppel-like factor 4, Gut-enriched krueppel-like factor, Kruppel-like factor 4 (gut) |
| Wirtsspezies | Mouse |
| Immunogen | This antibody was developed using a synthetic peptide derived from O43474, with the exact immunogen sequence remaining proprietary. |
| Reinigungsverfahren | Protein A purified |
| Mehr anzeigen |