missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KU70 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 10723-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
XRCC6 is a nuclear complex consisting of two subunits with molecular masses of approximately 70 and 80 kDa. The complex functions as a single-stranded DNA-dependent ATP-dependent helicase. The complex may be involved in the repair of nonhomologous DNA ends such as that required for double-strand break repair, transposition, and V(D)J recombination. High levels of autoantibodies to p70 and p80 have been found in some patients with systemic lupus erythematosus.Spezifikation
| KU70 | |
| Polyclonal | |
| Unconjugated | |
| XRCC6 | |
| 5 dRP lyase Ku70, 70 kDa subunit of Ku antigen, CTC75, CTCBF, DNA repair protein XRCC6, G22P1, KU70, ML8, Thyroid lupus autoantigen, TLAA, XRCC6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 25019, 2547 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P12956 | |
| Xrcc6 | |
| KU70 Fusion Protein Ag1043 | |
| 20 μL | |
| Primary | |
| Rat, Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur