missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-AP4, MedChemExpress
Beschreibung
L-AP4 (L-APB) is a potent and specific agonist for the group III mGluRs, with EC50s of 0.13, 0.29, 1.0, 249 μM for mGlu4, mGlu8, mGlu6 and mGlu7 receptors, respectively.
Spezifikation
Spezifikation
| Chemischer Name oder Material | L-AP4 |
| Physikalische Form | Powder |
| CAS | 23052-81-5 |
| Farbe | White |
| Menge | 5 mg |
| Hinweise zur Reinheitsqualität | Research |
| Summenformel | C4H10NO5P |
| Zur Verwendung mit (Anwendung) | Neuroscience-Neuromodulation |
| Synonym | L-APB |
| Löslichkeitsinformationen | H2O : 50 mg/mL (273.07 mM; Need ultrasonic) |
| Mehr anzeigen |
Research purposes only
Name des Produkts
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?