missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD23 is identical to low affinity IgE receptor found on B cells. CD23 is expressed on a subpopulation of peripheral blood cells, B lymphocytes and on EBV transformed B lymphoblastoid cell lines. CD23 is also detected in neoplastic cells from cases of B cell chronic lymphocytic leukemia and some cases on centroblastic/centrocytic lymphoma.
Host Species: Mouse
Clone: 1B12
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with rat. Others not tested.
Epitope: Extracellular
Immunogen: Recombinant external domain of CD23 protein
Molecular Weight: 45 kDa
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | CD23 Ab-1 |
| Anwendungen | Immunohistochemistry (Paraffin) |
| Klassifikation | Monoclonal |
| Klon | 1B12 |
| Konjugat | Unconjugated |
| Wirtsspezies | Mouse |
| Immunogen | Recombinant external domain of CD23 protein |
| Menge | 7 mL |
| Regulatorischer Status | IVD |
| Forschungsgebiet | Oncogenesis |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?