missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Helicobacter pylori is known to cause peptic ulcers and chronic gastritis in human. It is associated with duodenal ulcers and may be involved in development of adenocarcimona and low grade lymphoma of mucosa associated lymphoid tissue in the stomach.
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Helicobacter pylori
Epitope: Not determined
Immunogen: Purified Helicobacter pylori
Positive Control: Helicobacter pylori infected tissue
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | Helicobacter pylori |
| Anwendungen | Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Klon | Polyclonal |
| Konzentration | 200µg/mL |
| Konjugat | Unconjugated |
| Wirtsspezies | Rabbit |
| Immunogen | Purified Helicobacter pylori |
| Menge | 100 μL |
| Regulatorischer Status | IVD |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?