Learn More
Beschreibung
Host Species: Mouse
Clone: DM1A
Isotype: IgG1/κ
Species Reactivity: Human, Cow, Pig, Rat, Mouse, Guinea Pig, Gerbil, Frog and Chicken. Others not known.
Epitope: aa 426-450
Immunogen: Native chick brain microtubules
Molecular Weight: 57kDa
Positive Control: Ls174T, MAD109 cells; skin or lung
Cellular Localization: Cytoplasmic Ab-2 causes the 10nm filaments to collapse into large lateral aggregates collecting in the cell periphery or tight juxtanuclear caps. It does not block microtubule assembly. It does not inhibit polymerization or depolymerization of platelet tubulin in vitro. It blocks (by 70-80%) the ability of tubulin dimers (with GppNHp bound) to promote a stable inhibition of adenylyl cyclase.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native and denatured)
- Electron Microscopy
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Tubulin, alpha Ab-2 |
| Anwendungen | Immunoassay, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Klassifikation | Monoclonal |
| Klon | DM1A |
| Konzentration | 200μg/mL |
| Konjugat | Unconjugated |
| Wirtsspezies | Mouse |
| Immunogen | Native chick brain microtubules |
| Menge | 500 μL |
| Regulatorischer Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.